Learn More
Abnova™ Human USP1 Full-length ORF (NP_003359.3, 1 a.a. - 785 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007398-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the ubiquitin-specific processing (UBP) family of proteases that is a deubiquitinating enzyme (DUB) with His and Cys domains. This protein is located in the cytoplasm and cleaves the ubiquitin moiety from ubiquitin-fused precursors and ubiquitinylated proteins. The protein specifically deubiquitinates a protein in the Fanconi anemia (FA) DNA repair pathway. Alternate transcriptional splice variants have been characterized. [provided by RefSeq]
Sequence: MPGVIPSESNGLSRGSPSKKNRLSLKFFQKKETKRALDFTDSQENEEKASEYRASEIDQVVPAAQSSPINCEKRENLLPFVGLNNLGNTCYLNSILQVLYFCPGFKSGVKHLFNIISRKKEALKDEANQKDKGNCKEDSLASYELICSLQSLIISVEQLQASFLLNPEKYTDELATQPRRLLNTLRELNPMYEGYLQHDAQEVLQCILGNIQETCQLLKKEEVKNVAELPTKVEEIPHPKEEMNGINSIEMDSMRHSEDFKEKLPKGNGKRKSDTEFGNMKKKVKLSKEHQSLEENQRQTRSKRKATSDTLESPPKIIPKYISENESPRPSQKKSRVKINWLKSATKQPSILSKFCSLGKITTNQGVKGQSKENECDPEEDLGKCESDNTTNGCGLESPGNTVTPVNVNEVKPINKGEEQIGFELVEKLFQGQLVLRTRCLECESLTERREDFQDISVPVQEDELSKVEESSEISPEPKTEMKTLRWAISQFASVERIVGEDKYFCENCHHYTEAERSLLFDKMPEVITIHLKCFAASGLEFDCYGGGLSKINTPLLTPLKLSLEEWSTKPTNDSYGLFAVVMHSGITISSGHYTASVKVTDLNSLELDKGNFVVDQMCEIGKPEPLNEEEARGVVENYNDEEVSIRVGGNTQPSKVLNKKNVEAIGLLGGQKSKADYELYNKASNPDKVASTAFAENRNSETSDTTGTHESDRNKESSDQTGINISGFENKISYVVQSLKEYEGKWLLFDDSEVKVTEEKDFLNSLSPSTSPTSTPYLLFYKKLSpecifications
NP_003359.3 | |
Liquid | |
7398 | |
USP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPGVIPSESNGLSRGSPSKKNRLSLKFFQKKETKRALDFTDSQENEEKASEYRASEIDQVVPAAQSSPINCEKRENLLPFVGLNNLGNTCYLNSILQVLYFCPGFKSGVKHLFNIISRKKEALKDEANQKDKGNCKEDSLASYELICSLQSLIISVEQLQASFLLNPEKYTDELATQPRRLLNTLRELNPMYEGYLQHDAQEVLQCILGNIQETCQLLKKEEVKNVAELPTKVEEIPHPKEEMNGINSIEMDSMRHSEDFKEKLPKGNGKRKSDTEFGNMKKKVKLSKEHQSLEENQRQTRSKRKATSDTLESPPKIIPKYISENESPRPSQKKSRVKINWLKSATKQPSILSKFCSLGKITTNQGVKGQSKENECDPEEDLGKCESDNTTNGCGLESPGNTVTPVNVNEVKPINKGEEQIGFELVEKLFQGQLVLRTRCLECESLTERREDFQDISVPVQEDELSKVEESSEISPEPKTEMKTLRWAISQFASVERIVGEDKYFCENCHHYTEAERSLLFDKMPEVITIHLKCFAASGLEFDCYGGGLSKINTPLLTPLKLSLEEWSTKPTNDSYGLFAVVMHSGITISSGHYTASVKVTDLNSLELDKGNFVVDQMCEIGKPEPLNEEEARGVVENYNDEEVSIRVGGNTQPSKVLNKKNVEAIGLLGGQKSKADYELYNKASNPDKVASTAFAENRNSETSDTTGTHESDRNKESSDQTGINISGFENKISYVVQSLKEYEGKWLLFDDSEVKVTEEKDFLNSLSPSTSPTSTPYLLFYKKL | |
RUO | |
USP1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
114.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
UBP | |
USP1 | |
Yes | |
wheat germ expression system |