Learn More
Abnova™ Human USP28 Full-length ORF (AAH65928.1, 1 a.a. - 583 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00057646-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The ubiquitin-dependent protein degradation pathway is essential for proteolysis of intracellular proteins and peptides. Enzymes that remove ubiquitin from ubiquitin-conjugated peptides, like USP28, affect the fate and degradation of intracellular proteins and are essential for maintenance of cell-free ubiquitin pools (Valero et al., 2001).[supplied by OMIM]
Sequence: MTAELQQDDAAGAADGHGSSCQMLLNQLREITGIQDPSFLHEALKASNGDITQAVSLLTDERVKEPSQDTVATEPSEVEGSAANKEVLAKVIDLTHDNKDDLQAAIALSLLESPKIQADGRDLNRMHEATSAETKRSKRKRCEVWGENPNPNDWRRVDGWPVGLKNVGNTCWFSAVIQSLFQLPEFRRLVLSYSLPQNVLENCRSHTEKRNIMFMQELQYLFALMMGSNRKFVDPSAALDLLKGAFRSSEEQQQDVSEFTHKLLDWLEDAFQLAVNVNSPRNKSENPMVQLFYGTFLTEGVREGKPFCNNETFGQYPLQVNGYRNLDECLEGAMVEGDVELLPSDHSVKYGQERWFTKLPPVLTFELSRFEFNQSLGQPEKIHNKLEFPQIIYMDRYMYRSKELIRNKRECIRKLKEEIKILQQKLERYVKYGSGPARFPLPDMLKYVIEFASTKPASESCPPESDTHMTLPLSSVHCSVSDQTSKESTSTESSSQDVESTFSSPEDSLPKSKPLTSSRSSMEMPSQPAPRTVTDEEINFVKTCLQRWRSEIEQDIQDLKTCIASTTQTIEQMYCDPLLRQVESpecifications
AAH65928.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
92.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1515 | |
USP28 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
57646 | |
USP28 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTAELQQDDAAGAADGHGSSCQMLLNQLREITGIQDPSFLHEALKASNGDITQAVSLLTDERVKEPSQDTVATEPSEVEGSAANKEVLAKVIDLTHDNKDDLQAAIALSLLESPKIQADGRDLNRMHEATSAETKRSKRKRCEVWGENPNPNDWRRVDGWPVGLKNVGNTCWFSAVIQSLFQLPEFRRLVLSYSLPQNVLENCRSHTEKRNIMFMQELQYLFALMMGSNRKFVDPSAALDLLKGAFRSSEEQQQDVSEFTHKLLDWLEDAFQLAVNVNSPRNKSENPMVQLFYGTFLTEGVREGKPFCNNETFGQYPLQVNGYRNLDECLEGAMVEGDVELLPSDHSVKYGQERWFTKLPPVLTFELSRFEFNQSLGQPEKIHNKLEFPQIIYMDRYMYRSKELIRNKRECIRKLKEEIKILQQKLERYVKYGSGPARFPLPDMLKYVIEFASTKPASESCPPESDTHMTLPLSSVHCSVSDQTSKESTSTESSSQDVESTFSSPEDSLPKSKPLTSSRSSMEMPSQPAPRTVTDEEINFVKTCLQRWRSEIEQDIQDLKTCIASTTQTIEQMYCDPLLRQVE | |
RUO | |
USP28 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |