missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human UTRN Partial ORF (NP_009055, 3328 a.a. - 3433 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007402-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16147375

  • 6845.00 SEK / 25µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene shares both structural and functional similarities with the dystrophin gene. It contains an actin-binding N-terminus, a triple coiled-coil repeat central region, and a C-terminus that consists of protein-protein interaction motifs which interact with dystroglycan protein components. The protein encoded by this gene is located at the neuromuscular synapse and myotendinous junctions, where it participates in post-synaptic membrane maintenance and acetylcholine receptor clustering. Mouse studies suggest that this gene may serve as a functional substitute for the dystrophin gene and therefore, may serve as a potential therapeutic alternative to muscular dystrophy which is caused by mutations in the dystrophin gene. Alternative splicing of the utrophin gene has been described; however, the full-length nature of these variants has not yet been determined. [provided by RefSeq]

Sequence: LEARMQILEDHNKQLESQLHRLRQLLEQPESDSRINGVSPWASPQHSALSYSLDPDASGPQFHQAAGEDLLAPPHDTSTDLTEVMEQIHSTFPSCCPNVPSRPQAM
Specifications

Specifications

NP_009055
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.4kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LEARMQILEDHNKQLESQLHRLRQLLEQPESDSRINGVSPWASPQHSALSYSLDPDASGPQFHQAAGEDLLAPPHDTSTDLTEVMEQIHSTFPSCCPNVPSRPQAM
RUO
UTRN
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
7402
UTRN (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DMDL/DRP/DRP1/FLJ23678
UTRN
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers