missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human VARS Partial ORF (NP_006286, 994 a.a. - 1102 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_006286 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7407 |
Molecular Weight (g/mol) | 37.73kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16147385
|
Abnova™
H00007407-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16137385
|
Abnova™
H00007407-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. The protein encoded by this gene belongs to class-I aminoacyl-tRNA synthetase family and is located in the class III region of the major histocompatibility complex. [provided by RefSeq]
Sequence: AVRLSNQGFQAYDFPAVTTAQYSFWLYELCDVYLECLKPVLNGVDQVAAECARQTLYTCLDVGLRLLSPFMPFVTEELFQRLPRRMPQAPPSLCVTPYPEPSECSWKDPSpecifications
NP_006286 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
G7A/VARS1/VARS2 | |
VARS | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
7407 | |
VARS (Human) Recombinant Protein (Q01) | |
AVRLSNQGFQAYDFPAVTTAQYSFWLYELCDVYLECLKPVLNGVDQVAAECARQTLYTCLDVGLRLLSPFMPFVTEELFQRLPRRMPQAPPSLCVTPYPEPSECSWKDP | |
RUO | |
VARS | |
Wheat Germ (in vitro) | |
GST | |
Liquid |