missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human VAT1 Partial ORF (NP_006364, 294 a.a. - 392 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_006364 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10493 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16162646
|
Abnova™
H00010493-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16152646
|
Abnova™
H00010493-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Synaptic vesicles are responsible for regulating the storage and release of neurotransmitters in the nerve terminal. The protein encoded by this gene is an abundant integral membrane protein of cholinergic synaptic vesicles and is thought to be involved in vesicular transport. It belongs to the quinone oxidoreductase subfamily of zinc-containing alcohol dehydrogenase proteins. [provided by RefSeq]
Sequence: PKRNLMALARTWWNQFSVTALQLLQANRAVCGFHLGYLDGEVELVSGVVARLLALYNQGHIKPHIDSVWPFEKVADAMKQMQEKKNVGKVLLVPGPEKESpecifications
NP_006364 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20230/VATI | |
VAT1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10493 | |
VAT1 (Human) Recombinant Protein (Q01) | |
PKRNLMALARTWWNQFSVTALQLLQANRAVCGFHLGYLDGEVELVSGVVARLLALYNQGHIKPHIDSVWPFEKVADAMKQMQEKKNVGKVLLVPGPEKE | |
RUO | |
VAT1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |