Learn More
Abnova™ Human VNN2 Full-length ORF (NP_511043.1, 1 a.a. - 467 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_511043.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8875 |
Molecular Weight (g/mol) | 79.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16133032
|
Abnova™
H00008875-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16143032
|
Abnova™
H00008875-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene product is a member of the Vanin family of proteins which share extensive sequence similarity with each other, and also with biotinidase. The family includes secreted and membrane-associated proteins, a few of which have been reported to participate in hematopoietic cell trafficking. No biotinidase activity has been demonstrated for any of the vanin proteins, however, they possess pantetheinase activity, which may play a role in oxidative-stress response. The encoded protein is a GPI-anchored cell surface molecule that plays a role in transendothelial migration of neutrophils. This gene lies in close proximity to, and in same transcriptional orientation as two other vanin genes on chromosome 6q23-q24. Two transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]
Sequence: MNENIDILETAIKQAAEQGARIIVTPEDALYGWKFTRETVFPYLEDIPDPQVNWIPCQDPHRFGHTPVQARLSCLAKDNSIYVLANLGDKKPCNSRDSTCPPNGYFQYNTNVVYNTEGKLVARYHKYHLYSEPQFNVPEKPELVTFNTAFGRFGIFTCFDIFFYDPGVTLVKDFHVDTILFPTAWMNVLPLLTAIEFHSAWAMGMGVNLLVANTHHVSLNMTGSGIYAPNGPKVYHYDMKTELGKLLLSEVDSHPLSSLAYPTAVNWNAYATTIKPFPVQKNTFRGFISRDGFNFTELFENAGNLTVCQKELCCHLSYRMLQKEENEVYVLGAFTGLHGRRRREYWQVCTMLKCKTTNLTTCGRPVETASTRFEMFSLSGTFGTEYVFPEVLLTEIHLSPGKFEVLKDGRLVNKNGSSGPILTVSLFGRWYTKDSLYSSCGTSNSAITYLLIFILLMIIALQNIVMLSpecifications
NP_511043.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
79.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MNENIDILETAIKQAAEQGARIIVTPEDALYGWKFTRETVFPYLEDIPDPQVNWIPCQDPHRFGHTPVQARLSCLAKDNSIYVLANLGDKKPCNSRDSTCPPNGYFQYNTNVVYNTEGKLVARYHKYHLYSEPQFNVPEKPELVTFNTAFGRFGIFTCFDIFFYDPGVTLVKDFHVDTILFPTAWMNVLPLLTAIEFHSAWAMGMGVNLLVANTHHVSLNMTGSGIYAPNGPKVYHYDMKTELGKLLLSEVDSHPLSSLAYPTAVNWNAYATTIKPFPVQKNTFRGFISRDGFNFTELFENAGNLTVCQKELCCHLSYRMLQKEENEVYVLGAFTGLHGRRRREYWQVCTMLKCKTTNLTTCGRPVETASTRFEMFSLSGTFGTEYVFPEVLLTEIHLSPGKFEVLKDGRLVNKNGSSGPILTVSLFGRWYTKDSLYSSCGTSNSAITYLLIFILLMIIALQNIVML | |
RUO | |
VNN2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8875 | |
VNN2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FOAP-4/GPI-80 | |
VNN2 | |
Recombinant | |
wheat germ expression system |