Learn More
Abnova™ Human VPS33B Full-length ORF (AAH16445.1, 1 a.a. - 617 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH16445.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 26276 |
Molecular Weight (g/mol) | 97kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16188272
|
Abnova™
H00026276-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16178272
|
Abnova™
H00026276-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene is a member of the Sec-1 domain family, and encodes the human ortholog of rat Vps33b which is homologous to the yeast class C Vps33 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway. [provided by RefSeq]
Sequence: MAFPHRPDAPELPDFSMLKRLARDQLIYLLEQLPGKKDLFIEADLMSPLDRIANVSILKQHEVDKLYKVENKPALSSNEQLCFLVRPRIKNMRYIASLVNADKLAGRTRKYKVIFSPQKFYACEMVLEEEGIYGDVSCDEWAFSLLPLDVDLLSMELPEFFRDYFLEGDQRWINTVAQALHLLSTLYGPFPNCYGIGRCAKMAYELWRNLEEEEDGETKGRRPEIGHIFLLDRDVDFVTALCSQVVYEGLVDDTFRIKCGSVDFGPEVTSSDKSLKVLLNAEDKVFNEIRNEHFSNVFGFLSQKARNLQAQYDRRRGMDIKQMKNFVSQELKGLKQEHRLLSLHIGACESIMKKKTKQDFQELIKTEHALLEGFNIRESTSYIEEHIDRQVSPIESLRLMCLLSITENGLIPKDYRSLKTQYLQSYGPEHLLTFSNLRRAGLLTEQAPGDTLTAVESKVSKLVTDKAAGKITDAFSSLAKRSNFRAISKKLNLIPRVDGEYDLKVPRDMAYVFSGAYVPLSCRIIEQVLERRSWQGLDEVVRLLNCSDFAFTDMTKEDKASSESLRLILVVFLGGCTFSEISALRFLGREKGYRFIFLTTAVTNSARLMEAMSEVKASpecifications
AAH16445.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
97kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAFPHRPDAPELPDFSMLKRLARDQLIYLLEQLPGKKDLFIEADLMSPLDRIANVSILKQHEVDKLYKVENKPALSSNEQLCFLVRPRIKNMRYIASLVNADKLAGRTRKYKVIFSPQKFYACEMVLEEEGIYGDVSCDEWAFSLLPLDVDLLSMELPEFFRDYFLEGDQRWINTVAQALHLLSTLYGPFPNCYGIGRCAKMAYELWRNLEEEEDGETKGRRPEIGHIFLLDRDVDFVTALCSQVVYEGLVDDTFRIKCGSVDFGPEVTSSDKSLKVLLNAEDKVFNEIRNEHFSNVFGFLSQKARNLQAQYDRRRGMDIKQMKNFVSQELKGLKQEHRLLSLHIGACESIMKKKTKQDFQELIKTEHALLEGFNIRESTSYIEEHIDRQVSPIESLRLMCLLSITENGLIPKDYRSLKTQYLQSYGPEHLLTFSNLRRAGLLTEQAPGDTLTAVESKVSKLVTDKAAGKITDAFSSLAKRSNFRAISKKLNLIPRVDGEYDLKVPRDMAYVFSGAYVPLSCRIIEQVLERRSWQGLDEVVRLLNCSDFAFTDMTKEDKASSESLRLILVVFLGGCTFSEISALRFLGREKGYRFIFLTTAVTNSARLMEAMSEVKA | |
RUO | |
VPS33B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
26276 | |
VPS33B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ14848 | |
VPS33B | |
Recombinant | |
wheat germ expression system |