missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human VPS54 Full-length ORF (AAH41868.1, 1 a.a. - 824 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH41868.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 51542 |
Molecular Weight (g/mol) | 119.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16180123
|
Abnova™
H00051542-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16170123
|
Abnova™
H00051542-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes for a protein that in yeast forms part of a trimeric vacuolar-protein-sorting complex that is required for retrograde transport of proteins from prevacuoles to the late Golgi compartment. As in yeast, mammalian Vps54 proteins contain a coiled-coil region and dileucine motifs. Alternative splicing results in multiple transcript variants encoding different isoforms [provided by RefSeq]
Sequence: MLPTKNRIKREKIHERCKNICPPKDTFERTLLHTHDKSRTDLEQVPKIFMKPDFALDDSLTFNSVLPWSHFNTAGGKGNRDAASSKLLQEKLSHYLDIVEVNIAHQISLRSEAFFHAMTSQHELQDYLRKTSQAVKMLRDKIAQIDKVMCEGSLHILRLALTRNNCVKVYNKLKLMATVHQTQPTVQVLLSTSEFVGALDLIATTQEVLQQELQGIHSFRHLGSQLCELEKLIDKMMIAEFSTYSHSDLNRPLEDDCQVLEECVINKVSQTEEIDTDVVVKLADQMRMLNFPQWFDLLKDIFSKFTIFLQRVKATLNIIHSVVLSVLDKNQRTRELEEISQQKNAAKDNSLDTEVAYLIHEGMFISDAFGEGELTPIAVDTTSQRNASPNSEPCSSDSVSEPECTTDSSSSKEHTSSSAIPGGVDIMVSEDMKLTDSELGKLANNIQELLYSASDICHDRAVKFLMSRAKDGFLEKLNSMEFITLSRLMETFILDTEQICGRKSTSLLGALQSQAIKFVNRFHEERKTKLSLLLDNERWKQADVPAEFQDLVDSLSDGKIALPEKKSGATEERKPAEVLIVEGQQYAVVGTVLLLIRIILEYCQCVDNIPSVTTDMLTRLSDLLKYFNSRSCQLVLGAGALQVVGLKTITTKNLALSSRCLQLIVHYIPVIRAHFEARLPPKQYSMLRHFGHITKDYHDHIAEISAKLVAIMDSLFDKLLSKYEVKAPVPSACFRNICKQMTKMHEAIFDHLPEEQTQMLFLRINASYKLHLKKQLSHLNVINDGGPQNGLVTADVAFYTGNLQALKGLKDLDLNMAEIWEQKRSpecifications
AAH41868.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
119.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MLPTKNRIKREKIHERCKNICPPKDTFERTLLHTHDKSRTDLEQVPKIFMKPDFALDDSLTFNSVLPWSHFNTAGGKGNRDAASSKLLQEKLSHYLDIVEVNIAHQISLRSEAFFHAMTSQHELQDYLRKTSQAVKMLRDKIAQIDKVMCEGSLHILRLALTRNNCVKVYNKLKLMATVHQTQPTVQVLLSTSEFVGALDLIATTQEVLQQELQGIHSFRHLGSQLCELEKLIDKMMIAEFSTYSHSDLNRPLEDDCQVLEECVINKVSQTEEIDTDVVVKLADQMRMLNFPQWFDLLKDIFSKFTIFLQRVKATLNIIHSVVLSVLDKNQRTRELEEISQQKNAAKDNSLDTEVAYLIHEGMFISDAFGEGELTPIAVDTTSQRNASPNSEPCSSDSVSEPECTTDSSSSKEHTSSSAIPGGVDIMVSEDMKLTDSELGKLANNIQELLYSASDICHDRAVKFLMSRAKDGFLEKLNSMEFITLSRLMETFILDTEQICGRKSTSLLGALQSQAIKFVNRFHEERKTKLSLLLDNERWKQADVPAEFQDLVDSLSDGKIALPEKKSGATEERKPAEVLIVEGQQYAVVGTVLLLIRIILEYCQCVDNIPSVTTDMLTRLSDLLKYFNSRSCQLVLGAGALQVVGLKTITTKNLALSSRCLQLIVHYIPVIRAHFEARLPPKQYSMLRHFGHITKDYHDHIAEISAKLVAIMDSLFDKLLSKYEVKAPVPSACFRNICKQMTKMHEAIFDHLPEEQTQMLFLRINASYKLHLKKQLSHLNVINDGGPQNGLVTADVAFYTGNLQALKGLKDLDLNMAEIWEQKR | |
RUO | |
VPS54 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51542 | |
VPS54 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HCC8/SLP-8p/VPS54L/hVps54L | |
VPS54 | |
Recombinant | |
wheat germ expression system |