missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human WDR23 Full-length ORF (AAH67132.1, 1 a.a. - 546 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH67132.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 80344 |
Molecular Weight (g/mol) | 88.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16124983
|
Abnova™
H00080344-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16114983
|
Abnova™
H00080344-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a WD repeat-containing protein that interacts with the COP9 signalosome, a macromolecular complex that interacts with cullin-RING E3 ligases and regulates their activity by hydrolyzing cullin-Nedd8 conjugates. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq]
Sequence: MGSRNSSSAGSGSGDPSEGLPRRGAGLRRSEEEEEEDEDVDLAQVLAYLLRRGQVRLVQGGGAANLQFIQALLDSEEENDRAWDGRLGDRYNPPVDATPDTRELEFNEIKTQVELATGQLGLRRAAQKHSFPRMLHQRERGLCHRGSFSLGEQSRVISHFLPNDLGFTDSYSQKAFCGIYSKDGQIFMSACQDQTIRLYDCRYGRFHKFKSIKARDVGWSVLDVAFTPDGNHFLYSSWSDYIHICNIYGEGDTHTALDLRPDERRFAVFSIAVSSDGREVLGGANDGCLYVFDREQNRRTLQIESHEDDVNAVAFADISSQILFSGGDDAICKVWDRRTMREDDPKPVGALAGHQDGITFIDSKGDARYLISNSKDQTIKLWDIRRFSSREGMEASRQAATQQNWDYRWQQVPKKAWRKLKLPGDSSLMTYRGHGVLHTLIRCRFSPIHSTGQQFIYSGCSTGKVVVYDLLSGHIVKKLTNHKACVRDVSWHPFEEKIVSSSWDGNLRLWQYRQAEYFQDDMPESEECASAPAPVPQSSTPFSSPQSpecifications
AAH67132.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
88.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGSRNSSSAGSGSGDPSEGLPRRGAGLRRSEEEEEEDEDVDLAQVLAYLLRRGQVRLVQGGGAANLQFIQALLDSEEENDRAWDGRLGDRYNPPVDATPDTRELEFNEIKTQVELATGQLGLRRAAQKHSFPRMLHQRERGLCHRGSFSLGEQSRVISHFLPNDLGFTDSYSQKAFCGIYSKDGQIFMSACQDQTIRLYDCRYGRFHKFKSIKARDVGWSVLDVAFTPDGNHFLYSSWSDYIHICNIYGEGDTHTALDLRPDERRFAVFSIAVSSDGREVLGGANDGCLYVFDREQNRRTLQIESHEDDVNAVAFADISSQILFSGGDDAICKVWDRRTMREDDPKPVGALAGHQDGITFIDSKGDARYLISNSKDQTIKLWDIRRFSSREGMEASRQAATQQNWDYRWQQVPKKAWRKLKLPGDSSLMTYRGHGVLHTLIRCRFSPIHSTGQQFIYSGCSTGKVVVYDLLSGHIVKKLTNHKACVRDVSWHPFEEKIVSSSWDGNLRLWQYRQAEYFQDDMPESEECASAPAPVPQSSTPFSSPQ | |
RUO | |
WDR23 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
80344 | |
WDR23 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp779A1629/GL014/PRO2389 | |
WDR23 | |
Recombinant | |
wheat germ expression system |