Läs mer
R&D Systems™ Human WT1 (AAA61299.1) VersaClone cDNA
Specifikationer
Specifikationer
| Gen | WT1 |
| Tillträdesnummer | AAA61299.1 |
| Koncentration | 10 μg at 0.2 μg/μL |
| Formulering | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Infoga typ | 1363 bp |
| Gen-ID (Entrez) | 7490 |
| Gene Alias | AWT1, GUD, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33 |
| Innehåll och lagring | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Art | Human |
| Kvantitet | 10 μg |