missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human YBX1 Partial ORF (NP_004550, 51 a.a. - 139 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004904-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: DKKVIATKVLGTVKWFNVRNGYGFINRNDTKEDVFVHQTAIKKNNPRKYLRSVGDGETVEFDVVEGEKGAEAANVTGPGGVPVQGSKYASpecifications
NP_004550 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DKKVIATKVLGTVKWFNVRNGYGFINRNDTKEDVFVHQTAIKKNNPRKYLRSVGDGETVEFDVVEGEKGAEAANVTGPGGVPVQGSKYA | |
RUO | |
YBX1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
4904 | |
YBX1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BP-8/CSDA2/CSDB/DBPB/MDR-NF1/MGC104858/MGC110976/MGC117250/NSEP-1/NSEP1/YB-1/YB1 | |
YBX1 | |
Recombinant | |
wheat germ expression system |