missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZA20D2 Partial ORF (NP_005998.1, 42 a.a. - 117 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007763-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16177925

  • 3343.96 SEK / 10µg

Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: QNSGRMSPMGTASGSNSPTSDSASVQRADTSLNNCEGAAGSTSEKSRNVPVAALPVTQQMTEMSISREDKITTPKT
Specifications

Specifications

NP_005998.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.1kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QNSGRMSPMGTASGSNSPTSDSASVQRADTSLNNCEGAAGSTSEKSRNVPVAALPVTQQMTEMSISREDKITTPKT
RUO
ZFAND5
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
7763
ZA20D2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ZA20D2/ZFAND5A/ZNF216
ZFAND5
Yes
Videos
SDS
Documents
Certificates