missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZC3HAV1 Partial ORF (NP_078901.3, 601 a.a. - 699 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_078901.3 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 56829 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16128816
|
Abnova™
H00056829-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16138816
|
Abnova™
H00056829-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a CCCH-type zinc finger protein that is thought to prevent infection by retroviruses. Studies of the rat homolog indicate that the protein may primarily function to inhibit viral gene expression and induce an innate immunity to viral infection. Alternative splicing occurs at this locus and two variants, each encoding distinct isoforms, are described. [provided by RefSeq]
Sequence: SVFTTKWIWYWKNESGTWIQYGEEKDKRKNSNVDSSYLESLYQSCPRGVVPFQAGSRNYELSFQGMIQTNIASKTQKDVIRRPTFVPQWYVQQMKRGPESpecifications
NP_078901.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686F2052/DKFZp686H1869/DKFZp686O19171/FLB6421/FLJ13288/MGC48898/ZAP/ZC3H2/ZC3HDC2 | |
ZC3HAV1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
56829 | |
ZC3HAV1 (Human) Recombinant Protein (Q01) | |
SVFTTKWIWYWKNESGTWIQYGEEKDKRKNSNVDSSYLESLYQSCPRGVVPFQAGSRNYELSFQGMIQTNIASKTQKDVIRRPTFVPQWYVQQMKRGPE | |
RUO | |
ZC3HAV1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |