Learn More
Abnova™ Human ZFP95 Full-length ORF (AAH30790, 1 a.a. - 839 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023660-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a zinc finger protein of the Kruppel family. The protein contains a SCAN box and a KRAB A domain. A similar protein in mouse is differentially expressed in spermatogenesis. Two alternatively spliced transcript variants differing only in the 5' UTR have been described. Additional variants have been found, but their full-length sequences have not been determined. [provided by RefSeq]
Sequence: MIMTESREVIDLDPPAETSQEQEDLFIVKVEEEDCTWMQEYNPPTFETFYQRFRHFQYHEASGPREALSQLRVLCCEWLRPELHTKEQILELLVLEQFLTILPEEFQPWVREHHPESGEEAVAVIENIQRELEERRQQIVACPDVLPRKMATPGAVQESCSPHPLTVDTQPEQAPQKPRLLEENALPVLQVPSLPLKDSQELTASLLSTGSQKLVKIEEVADVAVSFILEEWGHLDQSQKSLYRDDRKENYGSITSMGYESRDNMELIVKQISDDSESHWVAPEHTERSVPQDPDFAEVSDLKGMVQRWQVNPTVGKSRQNPSQKRDLDAITDISPKQSTHGERGHRCSDCGKFFLQASNFIQHRRIHTGEKPFKCGECGKSYNQRVHLTQHQRVHTGEKPYKCQVCGKAFRVSSHLVQHHSVHSGERPYGCNECGKNFGRHSHLIEHLKRHFREKSQRCSDKRSKNTKLSVKKKISEYSEADMELSGKTQRNVSQVQDFGEGCEFQGKLDRKQGIPMKEILGQPSSKRMNYSEVPYVHKKSSTGERPHKCNECGKSFIQSAHLIQHQRIHTGEKPFRCEECGKSYNQRVHLTQHQRVHTGEKPYTCPLCGKAFRVRSHLVQHQSVHSGERPFKCNECGKGFGRRSHLAGHLRLHSREKSHQCRECGEIFFQYASLIEHQVLHMGQKNEKNGICEEAYSWNLTVIEDKKIELQEQPYQCDICGKAFGYSSDLIQHYRTHTAEKPYQCDICRENVGQCSHTKQHQKIYSSTKSHQCHECGRGFTLKSHLNQHQGIHTGEKPFQCKECGMNFSWSCSLFKHLRSHERTDPINTLSVEGSLLSpecifications
AAH30790 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
118.03kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ39233/KIAA1015/MGC33710/ZFP95 | |
ZKSCAN5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
23660 | |
ZFP95 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MIMTESREVIDLDPPAETSQEQEDLFIVKVEEEDCTWMQEYNPPTFETFYQRFRHFQYHEASGPREALSQLRVLCCEWLRPELHTKEQILELLVLEQFLTILPEEFQPWVREHHPESGEEAVAVIENIQRELEERRQQIVACPDVLPRKMATPGAVQESCSPHPLTVDTQPEQAPQKPRLLEENALPVLQVPSLPLKDSQELTASLLSTGSQKLVKIEEVADVAVSFILEEWGHLDQSQKSLYRDDRKENYGSITSMGYESRDNMELIVKQISDDSESHWVAPEHTERSVPQDPDFAEVSDLKGMVQRWQVNPTVGKSRQNPSQKRDLDAITDISPKQSTHGERGHRCSDCGKFFLQASNFIQHRRIHTGEKPFKCGECGKSYNQRVHLTQHQRVHTGEKPYKCQVCGKAFRVSSHLVQHHSVHSGERPYGCNECGKNFGRHSHLIEHLKRHFREKSQRCSDKRSKNTKLSVKKKISEYSEADMELSGKTQRNVSQVQDFGEGCEFQGKLDRKQGIPMKEILGQPSSKRMNYSEVPYVHKKSSTGERPHKCNECGKSFIQSAHLIQHQRIHTGEKPFRCEECGKSYNQRVHLTQHQRVHTGEKPYTCPLCGKAFRVRSHLVQHQSVHSGERPFKCNECGKGFGRRSHLAGHLRLHSREKSHQCRECGEIFFQYASLIEHQVLHMGQKNEKNGICEEAYSWNLTVIEDKKIELQEQPYQCDICGKAFGYSSDLIQHYRTHTAEKPYQCDICRENVGQCSHTKQHQKIYSSTKSHQCHECGRGFTLKSHLNQHQGIHTGEKPFQCKECGMNFSWSCSLFKHLRSHERTDPINTLSVEGSLL | |
RUO | |
ZKSCAN5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |