missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF154 Partial ORF (NP_003435, 1 a.a. - 98 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007710-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MAVATLRTPTQGTVTFEDVAVHFSWEEWGLLDEAQRCLYRDVMLENLALLTSLDVHHQKQHLGEKHFRSNVGRALFVKTCTFHVSGEPSTCREVGKDFSpecifications
NP_003435 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAVATLRTPTQGTVTFEDVAVHFSWEEWGLLDEAQRCLYRDVMLENLALLTSLDVHHQKQHLGEKHFRSNVGRALFVKTCTFHVSGEPSTCREVGKDF | |
RUO | |
ZNF154 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7710 | |
ZNF154 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC176628/MGC176661/pHZ-92 | |
ZNF154 | |
Recombinant | |
wheat germ expression system |