Learn More
Abnova™ Human ZNF165 Full-length ORF (AAH26092, 1 a.a. - 485 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007718-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the Kruppel family of zinc finger proteins. Members of this DNA-binding protein family act as transcriptional regulators. This gene is located within a cluster of zinc finger family members. The encoded protein may play a role in spermatogenesis. [provided by RefSeq]
Sequence: MATEPKKAAAQNSPEDEGLLIVKIEEEEFIHGQDTCLQRSELLKQELCRQLFRQFCYQDSPGPREALSRLRELCCQWLKPEIHTKEQILELLVLEQFLTILPGDLQAWVHEHYPESGEEAVTILEDLERGTDEAVLQVQAHEHGQEIFQKKVSPPGPALNVKLQPVETKAHFDSSEPQLLWDCDNESENSRSMPKLEIFEKIESQRIISGRISGYISEASGESQDICKSAGRVKRQWEKESGESQRLSSAQDEGFGKILTHKNTVRGEIISHDGCERRLNLNSNEFTHQKSCKHGTCDQSFKWNSDFINHQIIYAGEKNHQYGKSFKSPKLAKHAAVFSGDKTHQCNECGKAFRHSSKLARHQRIHTGERCYECNECGKSFAESSDLTRHRRIHTGERPFGCKECGRAFNLNSHLIRHQRIHTREKPYECSECGKTFRVSSHLIRHFRIHTGEKPYECSECGRAFSQSSNLSQHQRIHMRENLLMSpecifications
AAH26092 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
79.09kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LD65/ZSCAN7 | |
ZNF165 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7718 | |
ZNF165 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MATEPKKAAAQNSPEDEGLLIVKIEEEEFIHGQDTCLQRSELLKQELCRQLFRQFCYQDSPGPREALSRLRELCCQWLKPEIHTKEQILELLVLEQFLTILPGDLQAWVHEHYPESGEEAVTILEDLERGTDEAVLQVQAHEHGQEIFQKKVSPPGPALNVKLQPVETKAHFDSSEPQLLWDCDNESENSRSMPKLEIFEKIESQRIISGRISGYISEASGESQDICKSAGRVKRQWEKESGESQRLSSAQDEGFGKILTHKNTVRGEIISHDGCERRLNLNSNEFTHQKSCKHGTCDQSFKWNSDFINHQIIYAGEKNHQYGKSFKSPKLAKHAAVFSGDKTHQCNECGKAFRHSSKLARHQRIHTGERCYECNECGKSFAESSDLTRHRRIHTGERPFGCKECGRAFNLNSHLIRHQRIHTREKPYECSECGKTFRVSSHLIRHFRIHTGEKPYECSECGRAFSQSSNLSQHQRIHMRENLLM | |
RUO | |
ZNF165 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |