Learn More
Abnova™ Human ZNF180 Full-length ORF (AAH33642.1, 1 a.a. - 691 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007733-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Zinc finger proteins have been shown to interact with nucleic acids and to have diverse functions. The zinc finger domain is a conserved amino acid sequence motif containing 2 specifically positioned cysteines and 2 histidines that are involved in coordinating zinc. Kruppel-related proteins form 1 family of zinc finger proteins. See MIM 604749 for additional information on zinc finger proteins.[supplied by OMIM]
Sequence: MRRVYAGSWKRCAAQDLSTLLCLEESMEEQDEKPPEPPKACAQDSFLPQEIIIKVEGEDTGSLTIPSQEGVNFKIVTVDFTREEQGTWNPAQRTLDRDVILENHRDLVSWDLATAVGKKDSTSKQRIFDEEPANGVKIERFTRDDPWLSSCEEVDDCKDQLEKQQEKQEILLQEVAFTQRKAVIHERVCKSDETGEKSGLNSSLFSSPVIPIRNHFHKHVSHAKKWHLNAAVNSHQKINENETLYENNECGKPPQSIHLIQFTRTQTKDKCYGFSDRIQSFCHGTPLHIHEKIHGGGKTFDFKECGQVLNPKISHNEQQRIPFEESQYKCSETSHSSSLTQNMRNNSEEKPFECNQCGKSFSWSSHLVAHQRTHTGEKPYECSECGKSFSRSSHLVSHQRTHTGEKPYRCNQCGKSFSQSYVLVVHQRTHTGEKPYECNQCGKSFRQSYKLIAHQRTHTGEKPYECNQCGKSFIQSYKLIAHQRIHTGEKPYECNQCGKSFSQSYKLVAHQRTHTGEKPFECNQCGKSFSWSSQLVAHQRTHTGEKPYECSECGKSFNRSSHLVMHQRIHTGEKPYECNQCGKSFSQSYVLVVHQRTHTGEKPYECSQCGKSFRQSSCLTQHQRTHTGEKPFECNQCGKTFSLSARLIVHQRTHTGEKPFTCIQCGKAFINSYKLIRHQATHTEEKLYECNSpecifications
AAH33642.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
105.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HHZ168 | |
ZNF180 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7733 | |
ZNF180 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRRVYAGSWKRCAAQDLSTLLCLEESMEEQDEKPPEPPKACAQDSFLPQEIIIKVEGEDTGSLTIPSQEGVNFKIVTVDFTREEQGTWNPAQRTLDRDVILENHRDLVSWDLATAVGKKDSTSKQRIFDEEPANGVKIERFTRDDPWLSSCEEVDDCKDQLEKQQEKQEILLQEVAFTQRKAVIHERVCKSDETGEKSGLNSSLFSSPVIPIRNHFHKHVSHAKKWHLNAAVNSHQKINENETLYENNECGKPPQSIHLIQFTRTQTKDKCYGFSDRIQSFCHGTPLHIHEKIHGGGKTFDFKECGQVLNPKISHNEQQRIPFEESQYKCSETSHSSSLTQNMRNNSEEKPFECNQCGKSFSWSSHLVAHQRTHTGEKPYECSECGKSFSRSSHLVSHQRTHTGEKPYRCNQCGKSFSQSYVLVVHQRTHTGEKPYECNQCGKSFRQSYKLIAHQRTHTGEKPYECNQCGKSFIQSYKLIAHQRIHTGEKPYECNQCGKSFSQSYKLVAHQRTHTGEKPFECNQCGKSFSWSSQLVAHQRTHTGEKPYECSECGKSFNRSSHLVMHQRIHTGEKPYECNQCGKSFSQSYVLVVHQRTHTGEKPYECSQCGKSFRQSSCLTQHQRTHTGEKPFECNQCGKTFSLSARLIVHQRTHTGEKPFTCIQCGKAFINSYKLIRHQATHTEEKLYECN | |
RUO | |
ZNF180 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |