missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF224 Partial ORF (NP_037530.1, 95 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007767-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16117935

  • 6845.00 SEK / 25µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: HQEWSFQQIWEKIASDLTRSQDLVINSSQFSKEGDFPCQTEAGLSVIHTRQKSSQGNGYKPSFSDVSHFDFHQQLHSGEKSHTCDECGKNFCYISA
Specifications

Specifications

NP_037530.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.3kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HQEWSFQQIWEKIASDLTRSQDLVINSSQFSKEGDFPCQTEAGLSVIHTRQKSSQGNGYKPSFSDVSHFDFHQQLHSGEKSHTCDECGKNFCYISA
RUO
ZNF224
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
7767
ZNF224 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BMZF-2/BMZF2/KOX22/ZNF255/ZNF27
ZNF224
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers