Learn More
Abnova™ Human ZNF238 Partial ORF (NP_991331.1, 182 a.a. - 281 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010472-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
C2H2-type zinc finger proteins, such as ZNF238, act on the molecular level as transcriptional activators or repressors and are involved in chromatin assembly.[supplied by OMIM]
Sequence: KRDLAAEPGNMWMRLPSDSAGIPQAGGEAEPHATAAGKTVASPCSSTESLSQRSVTSVRDSADVDCVLDLSVKSSLSGVENLNSSYFSSQDVLRSNLVQVSpecifications
NP_991331.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KRDLAAEPGNMWMRLPSDSAGIPQAGGEAEPHATAAGKTVASPCSSTESLSQRSVTSVRDSADVDCVLDLSVKSSLSGVENLNSSYFSSQDVLRSNLVQV | |
RUO | |
ZNF238 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10472 | |
ZNF238 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C2H2-171/RP58/TAZ-1/ZBTB18 | |
ZNF238 | |
Recombinant | |
wheat germ expression system |