Learn More
Abnova™ Human ZNF385 Full-length ORF (NP_056296.1, 1 a.a. - 366 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00025946-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Zinc finger proteins, such as ZNF385A, are regulatory proteins that act as transcription factors, bind single- or double-stranded RNA, or interact with other proteins (Sharma et al., 2004 [PubMed 15527981]).[supplied by OMIM]
Sequence: MQPPLDLKQILPFPLEPAPTLGLFSNYSTMDPVQKAVLSHTFGGPLLKTKRPVISCNICQIRFNSQSQAEAHYKGNRHARRVKGIEAAKTRGREPGVREPGDPAPPGSTPTNGDGVAPRPVSMENGLGPAPGSPEKQPGSPSPPSIPETGQGVTKGEGGTPAPASLPGGSKEEEEKAKRLLYCALCKVAVNSLSQLEAHNKGTKHKTILEARSGLGPIKAYPRLGPPTPGEPEAPAQDRTFHCEICNVKVNSEVQLKQHISSRRHRDGVAGKPNPLLSRHKKSRGAGELAGTLTFSKELPKSLAGGLLPSPLAVAAVMAAAAGSPLSLRPAPAAPLLQGPPITHPLLHPAPGPIRTAHGPILFSPYSpecifications
NP_056296.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586G1122/HZF/RZF/ZFP385/ZNF385 | |
ZNF385A | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
25946 | |
ZNF385 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQPPLDLKQILPFPLEPAPTLGLFSNYSTMDPVQKAVLSHTFGGPLLKTKRPVISCNICQIRFNSQSQAEAHYKGNRHARRVKGIEAAKTRGREPGVREPGDPAPPGSTPTNGDGVAPRPVSMENGLGPAPGSPEKQPGSPSPPSIPETGQGVTKGEGGTPAPASLPGGSKEEEEKAKRLLYCALCKVAVNSLSQLEAHNKGTKHKTILEARSGLGPIKAYPRLGPPTPGEPEAPAQDRTFHCEICNVKVNSEVQLKQHISSRRHRDGVAGKPNPLLSRHKKSRGAGELAGTLTFSKELPKSLAGGLLPSPLAVAAVMAAAAGSPLSLRPAPAAPLLQGPPITHPLLHPAPGPIRTAHGPILFSPY | |
RUO | |
ZNF385A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |