missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF623 Full-length ORF (NP_055604.3, 1 a.a. - 536 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009831-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16134162

  • 4055.00 SEK / 10µg
Estimated Shipment: 17-11-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MILLSFVSDSNVGTGEKKVTEAWISEDENSHRTTSDRLTVMELPSPESEEVHEPRLGELLGNPEGQSLGSSPSQDRGCKQVTVTHWKIQTGETAQVCTKSGRNHILNSDLLLLQRELIEGEANPCDICGKTFTFNSDLVRHRISHAGEKPYTCDQCGKGFGQSSHLMEHQRIHTGERLYVCNVCGKDFIHYSGLIEHQRVHSGEKPFKCAQCGKAFCHSSDLIRHQRVHTRERPFECKECGKGFSQSSLLIRHQRIHTGERPYECNECGKSFIRSSSLIRHYQIHTEVKQYECKECGKAFRHRSDLIEHQRIHTGERPFECNECGKAFIRSSKLIQHQRIHTGERPYVCNECGKRFSQTSNFTQHQRIHTGEKLYECNECGKAFFLSSYLIRHQKIHTGERVYECKECGKAFLQKAHLTEHQKIHSGDRPFECKDCGKAFIQSSKLLLHQIIHTGEKPYVCSYCGKGFIQRSNFLQHQKIHTEEKLYECSQYGRDFNSTTNVKNNQRVHQEGLSLSKAPIHLGERSVDKGEHTGNL
Specifications

Specifications

NP_055604.3
Liquid
9831
ZNF623 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MILLSFVSDSNVGTGEKKVTEAWISEDENSHRTTSDRLTVMELPSPESEEVHEPRLGELLGNPEGQSLGSSPSQDRGCKQVTVTHWKIQTGETAQVCTKSGRNHILNSDLLLLQRELIEGEANPCDICGKTFTFNSDLVRHRISHAGEKPYTCDQCGKGFGQSSHLMEHQRIHTGERLYVCNVCGKDFIHYSGLIEHQRVHSGEKPFKCAQCGKAFCHSSDLIRHQRVHTRERPFECKECGKGFSQSSLLIRHQRIHTGERPYECNECGKSFIRSSSLIRHYQIHTEVKQYECKECGKAFRHRSDLIEHQRIHTGERPFECNECGKAFIRSSKLIQHQRIHTGERPYVCNECGKRFSQTSNFTQHQRIHTGEKLYECNECGKAFFLSSYLIRHQKIHTGERVYECKECGKAFLQKAHLTEHQKIHSGDRPFECKDCGKAFIQSSKLLLHQIIHTGEKPYVCSYCGKGFIQRSNFLQHQKIHTEEKLYECSQYGRDFNSTTNVKNNQRVHQEGLSLSKAPIHLGERSVDKGEHTGNL
RUO
ZNF623
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
87.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC103965/MGC104128
ZNF623
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers