Learn More
Abnova™ Human ZNRF1 Full-length ORF (NP_115644.1, 1 a.a. - 227 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00084937-P01.10ug
Additional Details : Weight : 0.00010kg
Description
In a study identifying genes in rat that are upregulated in response to nerve damage, a gene which is highly expressed in ganglia and in the central nervous system was found. The protein encoded by the rat gene contains both a zinc finger and a RING finger motif and is localized in the endosome/lysosome compartment, indicating that it may be involved in ubiquitin-mediated protein modification. The protein encoded by this human gene is highly similar in sequence to that encoded by the rat gene. [provided by RefSeq]
Sequence: MGGKQSTAARSRGPFPGVSTDDSAVPPPGGAPHFGHYRTGGGAMGLRSRSVSSVAGMGMDPSTAGGVPFGLYTPASRGTGDSERAPGGGGSASDSTYAHGNGYQETGGGHHRDGMLYLGSRASLADALPLHIAPRWFSSHSGFKCPICSKSVASDEMEMHFIMCLSKPRLSYNDDVLTKDAGECVICLEELLQGDTIARLPCLCIYHKSCIDSWFEVNRSCPEHPADSpecifications
NP_115644.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434E229/FLJ14846/MGC15430/NIN283 | |
ZNRF1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
84937 | |
ZNRF1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGGKQSTAARSRGPFPGVSTDDSAVPPPGGAPHFGHYRTGGGAMGLRSRSVSSVAGMGMDPSTAGGVPFGLYTPASRGTGDSERAPGGGGSASDSTYAHGNGYQETGGGHHRDGMLYLGSRASLADALPLHIAPRWFSSHSGFKCPICSKSVASDEMEMHFIMCLSKPRLSYNDDVLTKDAGECVICLEELLQGDTIARLPCLCIYHKSCIDSWFEVNRSCPEHPAD | |
RUO | |
ZNRF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |