missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZP2 Full-length ORF (AAH96305.1, 1 a.a. - 736 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007783-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16172112

  • 4515.00 SEK / 10µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The zona pellucida is an extracellular matrix that surrounds the oocyte and early embryo. It is composed primarily of three or four glycoproteins with various functions during fertilization and preimplantation development. The protein encoded by this gene is a structural component of the zona pellucida and functions in secondary binding and penetration of acrosome-reacted spermatozoa. The nascent protein contains a N-terminal signal peptide sequence, a conserved ZP domain, a consensus furin cleavage site, and a C-terminal transmembrane domain. It is hypothesized that furin cleavage results in release of the mature protein from the plasma membrane for subsequent incorporation into the zona pellucida matrix. However, the requirement for furin cleavage in this process remains controversial based on mouse studies. [provided by RefSeq]

Sequence: MACRQRGGSWSPSGWFNAGWSTYRSISLFFALVTSGNSIDVSQLVNPAFPGTVTCDEREITVEFPSSPGTKKWHASVVDPLGLDMPNCTYILDPEKLTLRATYDNCTRRVHGGHQMTIRVMNNSAALRHGAVMYQFFCPAMQVEETQGLSASTICQKDFMSFSLPRVFSGLADDSKGTKVQMGWSIEVGDGARAKTLTLPEAMKEGFSLLIDNHRMTFHVPFNATGVTHYVQGNSHLYMVSLKLTFISPGQKVIFSSQAICAPDPVTCNATHMTLTIPEFPGKLKSVSFENQNIDVSQLHDNGIDLEATNGMKLHFSKTLLKTKLSEKCLLHQFYLASLKLTFLLRPETVSMVIYPECLCESPVSIVTGELCTQDGFMDVEVYSYQTQPALDLGTLRVGNSSCQPVFEAQSQGLVRFHIPLNGCGTRYKFEDDKVVYENEIHALWTDFPPSKISRDSEFRNDMLLNINVESLTPPVASVKLGPFTLILQSYPDNSYQQPYGENEYPLVRFLRQPIYMEVRVLNRDDPNIKLVLDDCWATSTMDPDSFPQWNVVVDGCAYDLDNYQTTFHPVGSSVTHPDHYQRFDMKAFAFVSEAHVLSSLVYFHCSALICNRLSPDSPLCSVTCPVSSRHRRATGATEAEKMTVSLPGPILLLSDDSSFRGVGSSDLKASGSSGEKSRSETGEEVGSRGAMDTKGHKTAGDVGSKAVAAVAAFAGVVATLGFIYYLYEKRTVSNH
Specifications

Specifications

AAH96305.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
107.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ZPA
ZP2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
7783
ZP2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MACRQRGGSWSPSGWFNAGWSTYRSISLFFALVTSGNSIDVSQLVNPAFPGTVTCDEREITVEFPSSPGTKKWHASVVDPLGLDMPNCTYILDPEKLTLRATYDNCTRRVHGGHQMTIRVMNNSAALRHGAVMYQFFCPAMQVEETQGLSASTICQKDFMSFSLPRVFSGLADDSKGTKVQMGWSIEVGDGARAKTLTLPEAMKEGFSLLIDNHRMTFHVPFNATGVTHYVQGNSHLYMVSLKLTFISPGQKVIFSSQAICAPDPVTCNATHMTLTIPEFPGKLKSVSFENQNIDVSQLHDNGIDLEATNGMKLHFSKTLLKTKLSEKCLLHQFYLASLKLTFLLRPETVSMVIYPECLCESPVSIVTGELCTQDGFMDVEVYSYQTQPALDLGTLRVGNSSCQPVFEAQSQGLVRFHIPLNGCGTRYKFEDDKVVYENEIHALWTDFPPSKISRDSEFRNDMLLNINVESLTPPVASVKLGPFTLILQSYPDNSYQQPYGENEYPLVRFLRQPIYMEVRVLNRDDPNIKLVLDDCWATSTMDPDSFPQWNVVVDGCAYDLDNYQTTFHPVGSSVTHPDHYQRFDMKAFAFVSEAHVLSSLVYFHCSALICNRLSPDSPLCSVTCPVSSRHRRATGATEAEKMTVSLPGPILLLSDDSSFRGVGSSDLKASGSSGEKSRSETGEEVGSRGAMDTKGHKTAGDVGSKAVAAVAAFAGVVATLGFIYYLYEKRTVSNH
RUO
ZP2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers