Learn More
Abnova™ Human ZRF1 Full-length ORF (AAH56682.1, 1 a.a. - 167 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH56682.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 27000 |
Molecular Weight (g/mol) | 45.4kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16138462
|
Abnova™
H00027000-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16128462
|
Abnova™
H00027000-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is a member of the M-phase phosphoprotein (MPP) family. The gene encodes a phosphoprotein with a J domain and a Myb DNA-binding domain which localizes to both the nucleus and the cytosol. The protein is capable of forming a heterodimeric complex that associates with ribosomes, acting as a molecular chaperone for nascent polypeptide chains as they exit the ribosome. This protein was identified as a leukemia-associated antigen and expression of the gene is upregulated in leukemic blasts. Also, chromosomal aberrations involving this gene are associated with primary head and neck squamous cell tumors. This gene has a pseudogene on chromosome 6. Alternatively spliced variants which encode different protein isoforms have been described. [provided by RefSeq]
Sequence: MLLLPSAADGRGTAITHALTSASTLCQVEPVGRWFEAFVKRRNRNASASFQELEDKKELSEESEDEELQLEEFPMLKTLDPKDWKNQDHYAVLGLGHVRYKATQRQIKAAHKAMVLKHHPDKRKAAGEPIKEGDNDYFTCITKGSPGDSYQKLILQLIHLMKCYLIQSpecifications
AAH56682.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
MLLLPSAADGRGTAITHALTSASTLCQVEPVGRWFEAFVKRRNRNASASFQELEDKKELSEESEDEELQLEEFPMLKTLDPKDWKNQDHYAVLGLGHVRYKATQRQIKAAHKAMVLKHHPDKRKAAGEPIKEGDNDYFTCITKGSPGDSYQKLILQLIHLMKCYLIQ | |
RUO | |
DNAJC2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
27000 | |
ZRF1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MPHOSPH11/MPP11/ZRF1/ZUO1 | |
DNAJC2 | |
Recombinant | |
wheat germ expression system |