Läs mer
Proteintech™ HumanKine™ Recombinant Human FGF-4 Protein
Produktkod. 16881366 Handla allt Proteintech Produkter
Kvantitet:
10 μg
100μ g
1000 μg
Produktkod. 16881366
Brand: Proteintech™ HZ1218100UG
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
  • Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Specifikationer
Gen-ID (Entrez) 2249
Molekylvikt (g/mol) 17 and 27 kDa, monomer, glycosylated
Kvantitet 100μ g
Art Människa
Rekonstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS.
Innehåll och lagring Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Aktivitet Typically ≤ 1.25 ng/mL EC50
Uttryckssystem Human embryonic kidney 293 (HEK293) human cell expression system
Form Frystorkat material
Renhet eller kvalitetsklass > 95%