Läs mer
Proteintech™ HumanKine™ Recombinant Human FLT3 Ligand Protein
Produktkod. 16881336 Handla allt Proteintech Produkter
Kvantitet:
10 μg
100 μg
1000 μg
Produktkod. 16881336
Brand: Proteintech™ HZ11511000UG
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
  • Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Specifikationer
Gen-ID (Entrez) 2323
Molekylvikt (g/mol) 24 to 30 kDa, monomer, glycosylated
Kvantitet 1000 μg
Art Human
Rekonstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile water containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Innehåll och lagring Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Aktivitet Typically > 3 ng/mL EC50
Uttryckssystem Human embryonic kidney 293 (HEK293) human cell expression system
Form Lyophilized
Renhet eller kvalitetsklass > 95%