missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ HumanKine™ Recombinant Human FLT3 Ligand Protein
HumanKine™ cytokines and growth factors are human cell expressed proteins in HEK293 cells
1621.23 SEK - 31963.00 SEK
Specifications
Gene ID (Entrez) | 2323 |
---|---|
Molecular Weight (g/mol) | 24 to 30 kDa, monomer, glycosylated |
Species | Human |
Reconstitution | Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile water containing 0.1% endotoxin-free recombinant human serum albumin (HSA). |
Content And Storage | Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16801346
|
Proteintech™
HZ-1151-10UG |
10 μg |
1621.23 SEK
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16891336
|
Proteintech™
HZ-1151-100UG |
100 μg |
6378.40 SEK
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16881336
|
Proteintech™
HZ-1151-1000UG |
1000 μg |
31963.00 SEK
1000µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Specifications
2323 | |
Human | |
Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. | |
Human embryonic kidney 293 (HEK293) human cell expression system | |
> 95% |
24 to 30 kDa, monomer, glycosylated | |
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile water containing 0.1% endotoxin-free recombinant human serum albumin (HSA). | |
Typically > 3 ng/mL EC50 | |
Lyophilized |