Läs mer
Humulene, TRC
Produktkod. 17927454
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17927454 250 mg
17908004 100 mg
17998214 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17927454 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH674570250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6753-98-6
Molekylformel C15 H24
Molekylvikt (g/mol) 204.35
Synonym 1,4,8-Cycloundecatriene, 2,6,6,9-tetramethyl-, (1E,4E,8E)-, 1,4,8-Cycloundecatriene, 2,6,6,9-tetramethyl-, (E,E,E)- (8CI), α-Humulene (6CI,7CI), (1E,4E,8E)-2,6,6,9-Tetramethyl-1,4,8-cycloundecatriene, (±)-α-Humulene, Humulene, α-Caryophyllene
IUPAC-namn (1Z,4E,8E)-2,6,6,9-tetramethylcycloundeca-1,4,8-triene
LEDER C\C\1=C\CC\C(=C\CC(C)(C)\C=C\C1)\C
Specifikationer
Kemiskt namn eller material alpha-Humulene
Kvantitet 250 mg
InChI formel InChI=1S/C15H24/c1-13-7-5-8-14(2)10-12-15(3,4)11-6-9-13/h6-7,10-11H,5,8-9,12H2,1-4H3/b11-6+,13-7-,14-10+
Formel vikt 204.1878