missing translation for 'onlineSavingsMsg'
Learn More

HUS1B Rabbit anti-Human, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 18229-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16828294

  • 3313.85 SEK / 150µL
Estimated Shipment: 06-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

HUS1Bis most closely related to HUS1, a component of a cell cycle checkpoint protein complex involved in cell cycle arrest in response to DNA damage. HUS1B can interact with the check point protein RAD1 but not with RAD9. Overexpression of HUS1B has been shown to induce cell death, which suggests a related but distinct role of this protein, as compared to the HUS1.The protein encoded by this gene is most closely related to HUS1, a component of a cell cycle checkpoint protein complex involved in cell cycle arrest in response to DNA damage. This protein can interact with the check point protein RAD1 but not with RAD9. Overexpression of this protein has been shown to induce cell death, which suggests a related but distinct role of this protein, as compared to the HUS1.
Specifications

Specifications

HUS1B
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8NHY5
Checkpoint protein HUS1B, hHUS1B, HUS1B, RP11 532F6.1
Rabbit
IgG
150 μL
-20°C
Primary
135458, 691382
Western Blot
Unconjugated
HUS1B
Liquid
Hus1b
HUS1B Fusion Protein Ag12712
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers