missing translation for 'onlineSavingsMsg'
Learn More

Hyaluronic acid sodium salt, Streptococcus equi, 91%, Thermo Scientific Chemicals

Catalog Number p-7057577
Click to view available options
Quantity:
1 g
9067-32-7
(C14H20NO11Na)n
417.30
MFCD00875848
YWIVKILSMZOHHF-QJZPQSOGSA-N
Sodium hyaluronate
CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-*
9067-32-7
(C14H20NO11Na)n
417.30
MFCD00875848
YWIVKILSMZOHHF-QJZPQSOGSA-N
Sodium hyaluronate
CC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-*

Hyaluronic acid sodium salt, CAS # 9067-32-7, is an acidic mucopolysaccharide produced by the bacterium Streptococcus zooepidemicus that is abundant in the mammalian extracellular matrix.

Bacterial hyaluronic acid (HA) is a natural non-sulfated high molecular weight glycosaminoglycan that may be used to study formulations of non-animal gels potentially useful for procedures such as soft tissue augmentation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Hyaluronic acid sodium salt is a component of the extracellular matrix that is potentially useful for soft tissue augmentation procedures

Application

• Hyaluronic acid sodium salt may be used in formulations of non-animal gels used in soft tissue augmentation studies
• It can be used in laboratory development of artificial synovial fluid
• It can also be used to study its role in oxidative stress and tumor progression

TRUSTED_SUSTAINABILITY
Type Hyaluronic Acid Sodium Salt
Physical Form Powder
Merck Index 14,4757
Solubility Information Soluble in water
Percent Purity 91%
Color White
Quantity 1 g
Chemical Name or Material Hyaluronic acid sodium salt, Streptococcus equi

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'