Learn More
Axygen™ HybridRack™ Pipet Tip, Extended Length, Non-Filtered
The HybridRack System removes approximately 70% of the plastic compared to traditional plastic pipet tip racks.
711.62 SEK - 1009.73 SEK
Specifications
Filtered | Non-filtered |
---|---|
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Format | Rack |
Tip Style | Extended Length |
Product Code | Brand | Volume (Metric) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Color | Price | Quantity & Availability | ||||
30090436
![]() |
Axygen™
ER-10XT-R-S |
10 μL | Red |
711.62 SEK
Pack of 960 |
In Stock
Log in to see stock. ![]() |
||||
30090449
![]() |
Axygen™
ER-300XT-R-S |
300 μL | Green |
1009.73 SEK
Pack of 960 |
In Stock
Log in to see stock. ![]() |
||||
30090425
![]() |
Axygen™
ER-1000XT-R-S |
1000 μL | Blue |
1009.73 SEK
Pack of 960 |
In Stock
Log in to see stock. ![]() |
||||
Description
The Axygen HybridRack System combines sustainability and functionality to create a new, ecofriendly pipet tip product that is backed by Corning’s quality and value.
- The HybridRack System removes approximately 70% of the plastic compared to traditional plastic pipet tip racks.
- The plastic deck combined with the paperboard base provides a stable pipetting platform. The HybridRack is designed for end users to pipet directly from it, even when using a multi-channel instrument.
- The paperboard base can be easily recycled through most local curbside recycling programs.
- All HybridRack part numbers are provided sterile. Filtered, Maxymum Recovery, and non-filtered configurations are available.
- The HybridRack System is manufactured in the USA at an ISO 13485, ISO 14001, and EnergyStar registered facility.
- The pipet tips included in the HybridRack offering are the same tips as the Axygen MultiRack product line.
Specifications
Non-filtered | |
Not Autoclavable | |
Extended Length | |
Single-use | |
No |
Sterile | |
Rack | |
Pipet Tip | |
Universal Fit | |
Polypropylene |