Learn More
Axygen™ HybridRack™ Pipet Tip, Extended Length, Non-Filtered
GREENER_CHOICE
The HybridRack System removes approximately 70% of the plastic compared to traditional plastic pipet tip racks.
711.62 SEK - 1009.73 SEK
Specifications
Filtered Non-filtered
Sterility Sterile
Autoclavable Not Autoclavable
Format Rack
Tip Style Extended Length
View More Specs
Products 3
Product Code Brand Volume (Metric) Color Price Quantity & Availability  
Product Code Brand Volume (Metric) Color Price Quantity & Availability  
30090436
GREENER_CHOICE
View Documents Promotion Details
Axygen™
ER-10XT-R-S
10 μL Red
711.62 SEK
Pack of 960
Only null left
In Stock
Edge
Add to basket
30090449
GREENER_CHOICE
View Documents Promotion Details
Axygen™
ER-300XT-R-S
300 μL Green
1009.73 SEK
Pack of 960
Only null left
In Stock
Edge
Add to basket
30090425
GREENER_CHOICE
View Documents Promotion Details
Axygen™
ER-1000XT-R-S
1000 μL Blue
1009.73 SEK
Pack of 960
Only null left
In Stock
Edge
Add to basket
Description
Description
The Axygen HybridRack System combines sustainability and functionality to create a new, ecofriendly pipet tip product that is backed by Corning’s quality and value.
  • The HybridRack System removes approximately 70% of the plastic compared to traditional plastic pipet tip racks.
  • The plastic deck combined with the paperboard base provides a stable pipetting platform. The HybridRack is designed for end users to pipet directly from it, even when using a multi-channel instrument.
  • The paperboard base can be easily recycled through most local curbside recycling programs.
  • All HybridRack part numbers are provided sterile. Filtered, Maxymum Recovery, and non-filtered configurations are available.
  • The HybridRack System is manufactured in the USA at an ISO 13485, ISO 14001, and EnergyStar registered facility.
  • The pipet tips included in the HybridRack offering are the same tips as the Axygen MultiRack product line.
Specifications
Specifications
Non-filtered
Not Autoclavable
Extended Length
Single-use
No
Sterile
Rack
Pipet Tip
Universal Fit
Polypropylene