Learn More
Cytiva HyClone™ Hank's Balanced Salt Solution Powder
Available in lot sizes that facilitate scale-up and manufacturing.
360.00 SEK - 394.00 SEK
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
12329722
|
Cytiva
SH30016.02 |
Without Calcium, without Magnesium, with Phenol Red (powder) | 2 x 5 L |
374.00 SEK
Pack of 2 |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10474672
|
Cytiva
SH30015.03 |
With Calcium, with Magnesium, with Phenol Red (powder) | 1 x 10 L |
360.00 SEK
10 L |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11541801
|
Cytiva
SH30015.02 |
With Calcium, with Magnesium, with Phenol Red (powder) | 2 x 5 L |
394.00 SEK
Pack of 2 |
Estimated Shipment: 03-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
HyClone™ Hanks' Balanced Salt Solution Powder is available in lot sizes that facilitate scale-up and manufacturing.
Specifications
cGMP and ISO 9001:2000 Guidelines |