Learn More
HyClone™ HyPure Water for Injection (WFI) Quality Water, USP Specifications, Cytiva
Catalog Number p-7087592
Click to view available options
Quantity:
1 L
5 L
20 L
100 L
200 L
500 L
1000 L
Supports cell culture growth and biomanufacturing processes
  • Undergoes extensive purification to meet or exceed the stringent specifications of United States Pharmacopeia (USP)
  • Water for injection (WFI) is produced in ISO 9001 certified facilities to ensure a high quality product
  • Actual test results on a CoA are not taken from system samples, but must be from actual packaged samples of the given lot
  • Intended for further manufacturing or research use
  • Not for diagnostic or therapeutic use
Description Manufactured in an ISO9001 Certified Facility
Packaging Bottle
Filtered Through 0.1 µm Sterile Filter
Quantity 1 L
Physical Form Liquid
Product Line HyClone™