Läs mer
Hydrazinecarboximidothioic Acid Methyl Ester Hydroiodide, TRC
Produktkod. 30408217
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30408217 500 mg
1 options
Produktkod. 30408217 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH697568500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Hydrazinecarboximidothioic Acid Methyl Ester Hydroiodide
CAS 35600-34-1
Molekylformel C2H7N3S . (HI)
Kvantitet 500 mg
Synonym 3-Methylisothiosemicarbazide Hydriodide, S-Methylisothiosemicarbazide Hydriodide, S-Methylthiosemicarbazide Hydrogen Iodide
InChI formel InChI=1S/C2H7N3S.HI/c1-6-2(3)5-4;/h4H2,1H3,(H2,3,5);1H
LEDER I.CSC(=N)NN
IUPAC-namn methyl N-aminocarbamimidothioate;hydroiodide
Molekylvikt (g/mol) 233.08
Formel vikt 232.9484