missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrobromic Acid, 47% to 49%, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Description
- For Laboratory,Research,or Manufacturing Use
Specifications
Packaging | Narrow Mouth Amber Glass Bottle |
Specific Gravity | 1L = 1.50kg |
Quantity | 500 mL |
IUPAC Name | hydrogen bromide |
Formula Weight | 80.92 |
Percent Purity | 47 to 49% |
Grade | Meets ACS reagent chemical requirements |
Chemical Name or Material | Hydrobromic Acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'