missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrobromic Acid, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Concentration | 48% |
Density | 1.49g/mL at 25°C |
Residue after Ignition | ≤0.002% |
Vapor Pressure | 8mmHg (25°C), 320psi (21.1°C) |
Quantity | 100 mL |
UN Number | 1788 |
Beilstein | 3587158 |
IUPAC Name | hydrogen bromide |
Formula Weight | 80.91g/mol |
Grade | ACS Reagent |
Show More |
Safety and Handling
Product Identifier
- Hydrobromic acid
- Danger
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : hydrobromic acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'