missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic Acid Solution, Honeywell Fluka™

Catalog Number p-7071841
Click to view available options
Quantity:
50 mL
250 mL
1 L
This item is not returnable. View return policy
10035-10-6
BrH
80.91
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
260
CHEBI:47266
Br
This item is not returnable. View return policy
10035-10-6
BrH
80.91
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
260
CHEBI:47266
Br

33 wt. % in acetic acid

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Concentration 33wt. % in acetic acid
Density 1.40g/mL at 20°C
Flash Point >65°C (>149°F)
Quantity 50 mL
UN Number 3265
Beilstein 3587158
IUPAC Name hydrogen bromide
Formula Weight 80.91g/mol
Chemical Name or Material Hydrobromic Acid Solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'