Learn More
Hydrobromic Acid Solution, purum p.a., ≥62% (T), Honeywell™
Catalog Number 15631690
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass Bottle
10035-10-6
BrH
80.91
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
260
CHEBI:47266
Br
10035-10-6
BrH
80.91
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
260
CHEBI:47266
Br
Purum p.a.,≥62% (T)
  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY
Color Brown-Yellow
Melting Point -11°C
Boiling Point 126°C
Density 1.73g/cu.cm
Packaging Glass Bottle
Quantity 1 L
Physical Form Clear liquid
UN Number UN1788
Beilstein 3587158
IUPAC Name hydrogen bromide
Formula Weight 80.91g/mol
Percent Purity ≥62% (T)
Grade purum p.a.
Chemical Name or Material Hydrobromic Acid Solution