missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.04M, Chem-Lab

For laboratory use in Titrations - TMAR TA-TAC determination according to AFNOR T.90.036

Brand:  Chem Lab CL05.0332.9025

Additional Details : CAS Number : 7647-01-0 Weight : 25.00000kg Transport : UN number : 1789 Chem class : 8 Pack group : III

Quantity 25L

 View more versions of this product

Product Code. 11953143

  • 2735.00 SEK / 25 L

Estimated Shipment: 25-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
chlorane
ClH
MFCD00011324
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid
CHEBI:17883
[H+].[Cl-]

Specifications

Hydrochloric acid
7647-01-0
MFCD00011324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
1.4584g HCl / 1L H2O
1.00g/mL
313
36.46g/mol
0.04N (±0.00008 at 20°C)
25L
ClH
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid
[H+].[Cl-]
chlorane
36.46
CHEBI:17883
PE
Videos
SDS
Documents
Certificates