missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.1M, Chem Lab™

Brand:  Chem Lab CL05.0307.5500

Additional Details : CAS Number : 7647-01-0 Weight : 5.00000kg Transport : UN number : 1789 Chem class : 8 Pack group : III


Quantity 5L

 View more versions of this product

Product Code. 11953113

  • 397.00 SEK / 5 L

Estimated Shipment: 25-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

Specifications

5L
Colorless
3.646g HCl/l H2O = 0.1 N (±0.0002/20°C)
Liquid
<2
Keep container tightly closed.
100°C B.P., 0°C M.P.
Hydrochloric Acid 0.1mol/L
1g/mL
Odorless
Soluble in Water
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Hydrochloric acid 0.1 mol/l

Signal Word
  • Warning

Hazard Category
  • Substances and mixtures corrosive to metals Category 1

Hazard Statement
  • H290-May be corrosive to metals.

Precautionary Statement
  • P390-Absorb spillage to prevent material damage.

Supplemental information
  • MIXTURE LIST-Contains : HCl/H2O

SDS
Documents
Certificates