missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.1M, Chem-Lab

For laboratory use, Titrations

Brand:  Chem Lab CL05.0307.9520

Additional Details : CAS Number : 7647-01-0 Weight : 20.00000kg Transport : UN number : 1789 Chem class : 8 Pack group : III


Packaging EP
Quantity 20L

 View more versions of this product

Product Code. 11983113

  • 1510.00 SEK / 20 L

Estimated Shipment: 27-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
chlorane
ClH
MFCD00011324
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid
CHEBI:17883
[H+].[Cl-]

Specifications

Hydrochloric acid
7647-01-0
MFCD00011324
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid
[H+].[Cl-]
chlorane
36.46
CHEBI:17883
EP
20L
ClH
UN1789
VEXZGXHMUGYJMC-UHFFFAOYSA-N
3.646g HCl / 1L H2O
1.00g/mL
313
36.46g/mol
0.1N (±0.0002 at 20°C)
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Hydrochloric acid 0.1 mol/l

Signal Word
  • Warning

Hazard Category
  • Substances and mixtures corrosive to metals Category 1

Hazard Statement
  • H290-May be corrosive to metals.

Precautionary Statement
  • P390-Absorb spillage to prevent material damage.

Supplemental information
  • MIXTURE LIST-Contains : HCl/H2O

SDS
Documents
Certificates