missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.1M, Chem-Lab

Catalog Number p-7114735
Click to view available options
Quantity:
10 L
20 L
25 L
Packaging:
EP
PE
This item is not returnable. View return policy
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
CHEBI:17883
Cl
This item is not returnable. View return policy
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
CHEBI:17883
Cl

For laboratory use, Titrations

Concentration 3.646g HCl / 1L H2O
Density 1.00g/mL
Packaging PE
Normality 0.1N (±0.0002 at 20°C)
Quantity 10 L
UN Number UN1789
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid
IUPAC Name hydrogen chloride
Formula Weight 36.46g/mol
Chemical Name or Material Hydrochloric acid
compliance-icons
Product Identifier
  • Hydrochloric acid 0.1 mol/l
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contains : HCl/H2O
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'