missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 37%, ACS Reagent Grade, Scharlab

Catalog Number p-7083578
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
hydrogen chloride
Cl
This item is not returnable. View return policy
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
hydrogen chloride
Cl

Used as a laboratory reagent, acidifying agent, and in the production of chlorides and the synthesis of organic products.

  • Synonyms: Hydrochloric acid fuming, Muriatic acid, Hydrogen chloride solution
  • Molecular Formula: HCl
  • Molar Mass: 36.46g/mol
  • CAS: 7647-01-0
  • EIN ECS No.: 231-595-7
  • Residue After Evaporation: 0.001% max
Appearance of Solution Passes Test
Melting Point -28°C
Boiling Point ∼50°C
Density ∼1.19g/cm3
Identification Passes Test
Residue after Ignition 0.0003% max
Vapor Pressure 190hPa at 20°C
Quantity 1 L
Physical Form Solution
Solubility Information Miscible in water (20°C)
Formula Weight 36.46g/mol
Percent Purity 36.5 to 38.0% (acidimetric)
Grade ACS Reagent
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid, 37%, reagent grade, ACS, ISO, Reag. Ph Eur
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Hydrogen chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'