Learn More
Hydrochloric Acid Solution, 0.1M, Fisher Chemical™
BIOPHARMA
Product Code. 12970754 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
1000 mL
Unit Size:
1000mL
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12970754 100 mL 100mL
12900934 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12970754 Supplier Fisher Chemical Supplier No. SOL 113 100
  • ONLINE EXCLUSIVE PRICE
    519.00 SEK
    381.00 SEK / 100ml
    Save 138.00 SEK
    26% Av
 Restricted Item
Beräknad leverans: 21-12-2026
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Concentration 0.1mol/L
Packaging 100mL
Quantity 100 mL