Läs mer
Hydrocortisone 11,21-Diacetate, TRC
Produktkod. 30135668
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135668 250 mg
30133417 500 mg
30005174 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135668 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH714618250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 81968-66-3
Molekylformel C25 H34 O7
Molekylvikt (g/mol) 446.53
Synonym Pregn-4-ene-3,20-dione, 11,21-bis(acetyloxy)-17-hydroxy-, (11β)- (9CI), Cortisol, 11,21-diacetate (6CI,7CI), 17-Hydroxy-3,20-dioxopregn-4-ene-11β,21-diyl diacetate, Hydrocortisone 11,21-diacetate
IUPAC-namn [2-[(8S,9S,10R,11S,13S,14S,17R)-11-acetyloxy-17-hydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
LEDER CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@H](C[C@]12C)OC(=O)C
Specifikationer
Kemiskt namn eller material Hydrocortisone 11,21-Diacetate
Kvantitet 250 mg
InChI formel InChI=1S/C25H34O7/c1-14(26)31-13-21(29)25(30)10-8-19-18-6-5-16-11-17(28)7-9-23(16,3)22(18)20(32-15(2)27)12-24(19,25)4/h11,18-20,22,30H,5-10,12-13H2,1-4H3/t18-,19-,20-,22+,23-,24-,25-/m0/s1
Formel vikt 446.2305