Läs mer
Hydrocortisone 17,21-Methyl Orthobutyrate, TRC
Produktkod. 30408617
Change view
Click to view available options
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30408617 50 mg
30391655 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30408617 Supplier Toronto Research Chemicals Supplier No. TRCH71465550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13609-63-7
Molekylformel C26H38O6
Molekylvikt (g/mol) 446.58
Synonym Cortisol Cyclic 17,21-(Methyl Orthobutyrate), Orthobutyric Acid Methyl Ester Cyclic 17,21-Ester with Cortisol
IUPAC-namn (4R,8'S,9'S,10'R,11'S,13'S,14'S)-11'-hydroxy-2-methoxy-10',13'-dimethyl-2-propylspiro[1,3-dioxane-4,17'-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene]-3',5-dione
LEDER CCCC1(OC)OCC(=O)[C@]2(CC[C@H]3[C@@H]4CCC5=CC(=O)CC[C@]5(C)[C@H]4[C@@H](O)C[C@]23C)O1
Specifications
Kemiskt namn eller material Hydrocortisone 17,21-Methyl Orthobutyrate
Kvantitet 50 mg
InChI formel InChI=1S/C26H38O6/c1-5-10-26(30-4)31-15-21(29)25(32-26)12-9-19-18-7-6-16-13-17(27)8-11-23(16,2)22(18)20(28)14-24(19,25)3/h13,18-20,22,28H,5-12,14-15H2,1-4H3/t18-,19-,20-,22+,23-,24-,25-,26?/m0/s1
Formel vikt 446.2668