Läs mer
Hydrocortisone 17-Acetate, TRC
Produktkod. 30136213
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136213 500 mg
30040495 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136213 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH714695500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 16463-74-4
Molekylformel C23 H32 O6
Molekylvikt (g/mol) 404.5
Synonym Pregn-4-ene-3,20-dione, 17-(acetyloxy)-11,21-dihydroxy-, (11β)-, Cortisol, 17-acetate (7CI,8CI), (11β)-17-(Acetyloxy)-11,21-dihydroxypregn-4-ene-3,20-dione, Hydrocortisone 17-acetate
IUPAC-namn [(8S,9S,10R,11S,13S,14S,17R)-11-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
LEDER CC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)CO
Specifikationer
Kemiskt namn eller material Hydrocortisone 17-Acetate
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C23H32O6/c1-13(25)29-23(19(28)12-24)9-7-17-16-5-4-14-10-15(26)6-8-21(14,2)20(16)18(27)11-22(17,23)3/h10,16-18,20,24,27H,4-9,11-12H2,1-3H3/t16-,17-,18-,20+,21-,22-,23-/m0/s1
Formel vikt 404.2199
Procent renhet >95%