Läs mer
Hydrocortisone-17-hemisuccinate, TRC
Produktkod. 30012240
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012240 5 mg
30010804 25 mg
30007575 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012240 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH7165705MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 220489-73-6
Molekylformel C25H34O8
Molekylvikt (g/mol) 462.53
Synonym Pregn-4-ene-3,20-dione, 17-(3-carboxy-1-oxopropoxy)-11,21-dihydroxy-, (11β)- (9CI), Cortisol 17-hemisuccinate, Hydrocortisone 17-hemisuccinate
IUPAC-namn 4-[[(8S,9S,10R,11S,13S,14S,17R)-11-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]oxy]-4-oxo-butanoic acid
LEDER C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](OC(=O)CCC(=O)O)(C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23
Specifikationer
Kemiskt namn eller material Hydrocortisone-17-Succinate
Kvantitet 5 mg
InChI formel InChI=1S/C25H34O8/c1-23-9-7-15(27)11-14(23)3-4-16-17-8-10-25(19(29)13-26,33-21(32)6-5-20(30)31)24(17,2)12-18(28)22(16)23/h11,16-18,22,26,28H,3-10,12-13H2,1-2H3,(H,30,31)/t16-,17-,18-,22+,23-,24-,25-/m0/s1
Formel vikt 462.2254