Läs mer
Hydrocortisone 21-Aldehyde Hydrate, TRC
Produktkod. 30007365
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007365 25 mg
30006181 10 mg
30011744 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007365 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH71458025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 906337-64-2
Molekylformel C21H30O6
Molekylvikt (g/mol) 378.46
Synonym (11β)-11,17-Dihydroxy-3,20-dioxopregn-4-en-21-al, 11β,17-Dihydroxy-3,20- dioxopregn-4-en-21-al Hydrate, 11β,17-Dihydroxy-4-pregnene-3,20,21-trione Hydrate, 21-Dehydrocortisol, 21-Dehydrocortisol, Cortisol-21-aldehyde, Dehydrocortisol, NSC 15472, Hydrocortisone EP Impurity G
LEDER C[C@@]1([C@@]2(O)C(C([H])=O)=O)[C@](CC2)([H])[C@@](CCC3=CC4=O)([H])[C@]([C@]3(CC4)C)([H])[C@@H](O)C1>>C[C@@]5([C@@]6(O)C(C(O)O)=O)[C@](CC6)([H])[C@@](CCC7=CC8=O)([H])[C@]([C@]7(CC8)C)([H])[C@@H](O)C5
Specifikationer
Kemiskt namn eller material Hydrocortisone 21-Aldehyde Hydrate
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C21H28O5.H2O/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19;/h9,11,14-16,18,24,26H,3-8,10H2,1-2H3;1H2/t14-,15-,16-,18+,19-,20-,21-;/m0./s1
Procent renhet >95%