Läs mer
Hydrocortisone 21-Butyrate, TRC
Produktkod. 30132446
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132446 50 mg
30136555 100 mg
17903725 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132446 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH71462550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6677-99-2
Molekylformel C25 H36 O6
Molekylvikt (g/mol) 432.55
Synonym Hydrocortisone 21-butyrate, (11β)-11,17-Dihydroxy-21-(1-oxobutoxy)-pregn-4-ene-3,20-dione, Cortisol 21-butyrate, Butyric acid, 21-ester with cortisol
IUPAC-namn [2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] butanoate
LEDER CCCC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C
Specifikationer
Kemiskt namn eller material Hydrocortisone 21-Butyrate
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C25H36O6/c1-4-5-21(29)31-14-20(28)25(30)11-9-18-17-7-6-15-12-16(26)8-10-23(15,2)22(17)19(27)13-24(18,25)3/h12,17-19,22,27,30H,4-11,13-14H2,1-3H3/t17-,18-,19-,22+,23-,24-,25-/m0/s1
Formel vikt 432.2512
Procent renhet >95%